Anti-Capillary Morphogenesis Gene 2 Goat Polyclonal Antibody
Catalog # USBIC1077-45
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Capillary Morphogenesis Gene 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: Capillary Morphogenesis Gene 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse