Anti-Calcitonin Gene-related Peptide Type 1 Receptor Rabbit Polyclonal Antibody
USBIC0115-45A
- Antibody type:Primary
- Antigen name:Calcitonin Gene-related Peptide Type 1 Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:436 to 461
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: Calcitonin Gene-related Peptide Type 1 Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human