Anti-Calcitonin Gene Related Peptide 2 Rabbit polyclonal antibody unconjugated
USBIC0115-03C
- Antibody type:Primary
- Antigen name:Calcitonin Gene Related Peptide 2
- Antigen symbol:CGRP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Format:Serum
- Form:lyophilized powder
- Sequence:ACNTATCVTHRLAGLLSRSGGMVKSNFVPTNVGSKAF-NH2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Recommended dilution: Immunofluorescence: 1:200; Immunochemistry: 1:500
Type: Primary
Antigen: Calcitonin Gene Related Peptide 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat