Anti-Calcitonin Gene Related Peptide Rabbit polyclonal antibody unconjugated
USBIC0115-02J
- Antibody type:Primary
- Antigen name:Calcitonin Gene Related Peptide
- Antigen symbol:CGRP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Format:Serum
- Form:lyophilized powder
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Recommended dilution: Immunochemistry: 1:200
Type: Primary
Antigen: Calcitonin Gene Related Peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat