Anti-Calcitonin Gene Related Peptide, alpha Mouse Monoclonal Antibody [clone: 13A93]
USBIC0115-01H
- Antibody type:Primary
- Antigen name:Calcitonin Gene Related Peptide alpha
- Antigen symbol:CGRPA
- Clonality:Monoclonal
- Clone:13A93
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:0,01M phosphate buffer, pH 7,4, 0,5M NaCland 15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: Calcitonin Gene Related Peptide, alpha
Clonality: Monoclonal
Clone: 13A93
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human