Anti-Cadherin M Mouse monoclonal antibody unconjugated [clone: 9L427]
Catalog # USBIC0108-34Z1
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cadherin M
- Antigen symbol:CDHM
- Clonality:Monoclonal
- Clone:9L427
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:2X PBS, PEG, sucrose, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by thiophilic adsorption and size exclusion chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 5 ?g/ml; ELISA: 0. 05 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: Cadherin M
Clonality: Monoclonal
Clone: 9L427
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human