Anti-BCL2L13 Rabbit polyclonal antibody unconjugated
Catalog # USBIB0807-21D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:BCL2-like 13 (apoptosis facilitator)
- Antigen symbol:BCL2L13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: BCL2L13 (BCL2-like 13 (apoptosis facilitator))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human