Order Entry
Sweden
ContactUsLinkComponent
Anti-BAX Mouse monoclonal antibody unconjugated [clone: 6A7]
Anti-BAX Mouse monoclonal antibody unconjugated [clone: 6A7]
Catalog # USBIB0175-08A
Supplier:  US Biological
Anti-BAX Mouse monoclonal antibody unconjugated [clone: 6A7]
Catalog # USBIB0175-08A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BCL2-associated X Protein
  • Antigen symbol:
    BAX
  • Clonality:
    Monoclonal
  • Clone:
    6A7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    100mM borate buffered saline, pH 8.0. No oramine- containing buffer salts .
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: ? 1 ?g/ml

Type: Primary
Antigen: BAX (BCL2-associated X protein)
Clonality: monoclonal
Clone: 6A7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat