Anti-ATRIP Rabbit Polyclonal Antibody
Catalog # USBIA4154-11
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ATR interacting Protein
- Antigen symbol:ATRIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 182
- Storage buffer:PBS containing 10% glycerol and 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~3,8 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.75 ?g/ml; Immunoprecipitation: 0.1-0.5 ?g
Type: Primary
Antigen: ATRIP (ATR interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human