Anti-AP2A2 Rabbit Polyclonal Antibody (Biotin)
USBIA2298-51K-BIOT
- Antibody type:Primary
- Antigen name:adaptor-related protein complex 2, alpha 2 subunit
- Antigen symbol:AP2A2
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: AP2A2 (adaptor-related protein complex 2, alpha 2 subunit)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human