Anti-Amylase Rabbit Polyclonal Antibody
Katalog # USBIA2275-49A
Leverantör: US Biological
Specifikationer
- Antibody type:Primary
- Antigen name:Amylase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2, dextran. 500ul sterile ddH2O.Dilute further
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:1 mg
- Pk:1 mg
Specifikationer
Om denna produkt
Type: Primary
Antigen: Amylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: