Order Entry
Sweden
ContactUsLinkComponent
Anti-ADAMTSL2 Goat Polyclonal Antibody
Anti-ADAMTSL2 Goat Polyclonal Antibody
  USBIA0859-58M
Anti-ADAMTSL2 Goat Polyclonal Antibody
  USBIA0859-58M

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    ADAMTS-like 2
  • Antigensymbol:
    ADAMTSL2
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Goat
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Form:
    liquid
  • Storage buffer:
    Tris saline, pH 7,2, 0,5% BSA, 0,02% sodium azide.
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Rening:
    purified by peptide affinity chromatography
  • Storlek:
    100 µg
  • Förp.:
    100 µG

 

 

Recommended dilution: ELISA: 1:128000

Type: Primary
Antigen: ADAMTSL2 (ADAMTS-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human