Anti-MOAP1 Rabbit Polyclonal Antibody
Catalog # USBI145999
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:modulator of apoptosis 1
- Antigen symbol:MOAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-4?g/ml
Type: Primary
Antigen: MOAP1 (modulator of apoptosis 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse