Anti-NCLN Sheep Polyclonal Antibody
USBI145550
- Antibody type:Primary
- Antigen name:nicalin
- Antigen symbol:NCLN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:1 to 522
- Storage buffer:PBS,Trehalose.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 0.5 ?g/ml
Type: Primary
Antigen: NCLN (nicalin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat