Anti-NAALADL2 Mouse Monoclonal Antibody [clone: 817227]
Catalog # USBI145539
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:N-acetylated alpha-linked acidic dipeptidase-like 2
- Antigen symbol:NAALADL2
- Clonality:Monoclonal
- Clone:817227
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Form:lyophilized powder
- Storage buffer:PBS,Trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 825 ?g/ml
Type: Primary
Antigen: NAALADL2 (N-acetylated alpha-linked acidic dipeptidase-like 2)
Clonality: monoclonal
Clone: 817227
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human