Anti-GAP43 Mouse Monoclonal Antibody [clone: GAP-8A12]
Catalog # USBI144562
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Growth associated protein 43
- Antigen symbol:GAP43
- Clonality:Monoclonal
- Clone:GAP-8A12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Format:ascites
- Form:lyophilized powder
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: GAP43 (growth associated protein 43)
Clonality: monoclonal
Clone: GAP-8A12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat