Anti-CAMK4 Mouse Monoclonal Antibody [clone: 13k21]
USBI143148
- Antibody type:Primary
- Antigen name:calcium/calmodulin-dependent Protein kinase IV
- Antigen symbol:CAMK4
- Clonality:Monoclonal
- Clone:13k21
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:0,1M Tris-glycine, pH 7,4, 150mM NaCl, 0,2% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: CAMK4 (calcium/calmodulin-dependent protein kinase IV)
Clonality: monoclonal
Clone: 13k21
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human