Anti-Gamma Globulin, theta Mouse Polyclonal Antibody
Catalog # USBI137984
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Gamma Globulin, theta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Mouse
- Format:Serum
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:2 mg
- Pk:2 mg
Specifications
About this item
Type: Primary
Antigen: Gamma Globulin, theta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse