Anti-FLT4 Mouse Monoclonal Antibody [clone: KLT9]
Catalog # USBI136982
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fms-related tyrosine kinase 4
- Antigen symbol:FLT4
- Clonality:Monoclonal
- Clone:KLT9
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Format:Supernatant
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Immunochemistry: 1:50-1:100
Type: Primary
Antigen: FLT4 (fms-related tyrosine kinase 4)
Clonality: monoclonal
Clone: KLT9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human