Anti-KIAA1324 Rabbit Polyclonal Antibody
USBI136210
- Antibody type:Primary
- Antigen symbol:KIAA1324
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: KIAA1324 (KIAA1324)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human