Order Entry
Sweden
ContactUsLinkComponent
Anti-ZNF624 Mouse Polyclonal Antibody
Anti-ZNF624 Mouse Polyclonal Antibody
Catalog # USBI135781
Supplier:  US Biological
Anti-ZNF624 Mouse Polyclonal Antibody
Catalog # USBI135781
Supplier:  US Biological

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    zinc finger protein 624
  • Antigensymbol:
    ZNF624
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Mouse
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Rening:
    purified by Protein A chromatography
  • Storlek:
    50 µg
  • Förp.:
    50 µG

Specifications

About this item

Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: ZNF624 (zinc finger protein 624)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse