Anti-WARS2 Mouse Monoclonal Antibody [clone: 3D8]
Catalog # USBI135320
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:tryptophanyl tRNA synthetase 2, mitochondrial
- Antigen symbol:WARS2
- Clonality:Monoclonal
- Clone:3D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Amino acid number:1 to 90
- Storage buffer:PBS, pH 7,2.
- Sequence:MALHSMRKARERWSFIRALHKGSAAAPALQKDSKKRVFSGIQPTGILHLGNYLGAIESWVRLQDEYDSVLYSIVDLHSITVPQDPAVLRQ
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: WARS2 (tryptophanyl tRNA synthetase 2, mitochondrial)
Clonality: monoclonal
Clone: 3D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human