Anti-TSNAX Mouse Monoclonal Antibody [clone: 4D5]
USBI134816
- Antibody type:Primary
- Antigen name:translin-associated factor X
- Antigen symbol:TSNAX
- Clonality:Monoclonal
- Clone:4D5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgM kappa
- Reactivity:Human
- Western blot:Yes
- Format:ascites
- Form:liquid
- Amino acid number:191 to 290
- Storage buffer:ascites fluid.
- Sequence:VADLTGELMRMCINSVGNGDIDTPFEVSQFLRQVYDGFSFIGNTGPYEVSKKLYTLKQSLAKVENACYALKVRGSEIPKHMLADVFSVKTEMIDQEEGIS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: TSNAX (translin-associated factor X)
Clonality: monoclonal
Clone: 4D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human