Anti-TNFSF18 Mouse Monoclonal Antibody [clone: 6F7]
Catalog # USBI134560
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor (ligand) superfamily, member 18
- Antigen symbol:TNFSF18
- Clonality:Monoclonal
- Clone:6F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:68 to 178
- Storage buffer:PBS, pH 7,2.
- Sequence:KWQMASSEPPCVNKVSDWKLEILQNGLYLIYGQVAPNANYNDVAPFEVRLYKNKDMIQTLTNKSKIQNVGGTYELHVGDTIDLIFNSEHQVLKNNTYWGIILLANPQFIS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TNFSF18 (tumor necrosis factor (ligand) superfamily, member 18)
Clonality: monoclonal
Clone: 6F7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human