Anti-PRMT6 Rabbit Polyclonal Antibody
USBI131839
- Antikroppstyp:Primär
- Antikroppsnamn:Protein arginine methyltransferase 6
- Antigensymbol:PRMT6
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Immunutfällning:Yes
- Isotyp:IgG
- Reaktivitet:Human
- Western blot:Yes
- Mått:Serum
- Form:liquid
- Aminosyretal:1 to 316
- Sequence:MIADRVRTDAYRLGILRNWAALRGKTVLDVGAGTGILSIFCAQAGARRVYAVEASAIWQQAREVVRFNGLEDRVHVLPGPVETVELPEQVDAIVSEWMGYGLLHESMLSSVLHARTKWLKEGGLLLPASAELFIAPISDQMLEWRLGFWSQVKQHYGVDMSCLEGFATRCLMGHSEIVVQGLSGEDVLARPQRFAQLELSRAGLEQELEAGVGGRFRCSCYGSAPMHGFAIWFQVTFPGGESEKPLVLSTSPFHPATHWKQALLYLNEPVQVEQDTDVSGEITLLPSRDNPRRLRVLLRYKVGDQEEKTKDFAMED
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:100 µl
- Förp.:100 µl
Type: Primary
Antigen: PRMT6 (protein arginine methyltransferase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human