Anti-POLR3D Mouse Monoclonal Antibody [clone: 4E6]
USBI131598
- Antibody type:Primary
- Antigen name:Polymerase (RNA) III (DNA directed) polypeptide D (44kDa)
- Antigen symbol:POLR3D
- Clonality:Monoclonal
- Clone:4E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Amino acid number:296 to 395
- Storage buffer:PBS, pH 7,2.
- Sequence:GQVVLIKQEKDREAKLAENACTLADLTEGQVGKLLIRKSGRVQLLLGKVTLDVTMGTACSFLQELVSVGLGDSRTGEMTVLGHVKHKLVCSPDFESLLDH
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: POLR3D (polymerase (RNA) III (DNA directed) polypeptide D, 44kDa)
Clonality: monoclonal
Clone: 4000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human