Anti-PNN Mouse Monoclonal Antibody [clone: 2B4]
USBI131542
- Antibody type:Primary
- Antigen name:Pinin
- Antigen symbol:PNN
- Clonality:Monoclonal
- Clone:2B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Form:liquid
- Amino acid number:201 to 301
- Storage buffer:PBS, pH 7,2.
- Sequence:AKQTELRLLEQKVELAQLQEEWNEHNAKIIKYIRTKTKPHLFYIPGRMCPATQKLIEESQRKMNALFEGRRIEFAEQINKMEARPRRQSMKEKEHQVVRN
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: PNN (pinin, desmosome associated protein)
Clonality: monoclonal
Clone: 2B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Rat