Order Entry
Sweden
ContactUsLinkComponent
Anti-NOG Rabbit Polyclonal Antibody
Anti-NOG Rabbit Polyclonal Antibody
Catalog # USBI130457
Supplier:  US Biological
Anti-NOG Rabbit Polyclonal Antibody
Catalog # USBI130457
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Noggin
  • Antigen symbol:
    NOG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Serum
  • Form:
    liquid
  • Amino acid number:
    1 to 232
  • Sequence:
    MERCPSLGVTLYALVVVLGLRATPAGGQHYLHIRPAPSDNLPLVDLIEHPDPIFDPKEKDLNETLLRSLLGGHYDPGFMATSPPEDRPGGGGGAAGGAEDLAELDQLLRQRPSGAMPSEIKGLEFSEGLAQGKKQRLSKKLRRKLQMWLWSQTFCPVLYAWNDLGSRFWPRYVKVGSCFSKRSCSVPEGMVCKPSKSVHLTVLRWRCQRRGGQRCGWIPIQYPIISECKCSC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: NOG (noggin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human