Anti-NAV2 Mouse Monoclonal Antibody [clone: 4D11]
USBI130163
- Antibody type:Primary
- Antigen name:neuron navigator 2
- Antigen symbol:NAV2
- Clonality:Monoclonal
- Clone:4D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1878 to 1974
- Storage buffer:PBS, pH 7,2.
- Sequence:TGSTPLLRNSHSNSLISECMDSEAETVMQLRNELRDKEMKLTDIRLEALSSAHQLDQLREAMNRMQSEIEKLKAENDRLKSESQGSGCSRAPSQVS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: NAV2 (neuron navigator 2)
Clonality: monoclonal
Clone: 4D11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human