Anti-MSX1 Mouse Monoclonal Antibody [clone: 1D2]
USBI129943
- Antibody type:Primary
- Antigen name:msh homeobox 1
- Antigen symbol:MSX1
- Clonality:Monoclonal
- Clone:1D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:216 to 297
- Storage buffer:PBS, pH 7,2.
- Sequence:NRRAKAKRLQEAELEKLKMAAKPMLPPAAFGLSFPLGGPAAVAAAAGASLYGASGPFQRAALPVAPVGLYTAHVGYSMYHLT
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: MSX1 (msh homeobox 1)
Clonality: monoclonal
Clone: 1D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human