Anti-KLF15 Mouse Monoclonal Antibody [clone: 1F3]
Catalog # USBI128852
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Kruppel-like factor 15
- Antigen symbol:KLF15
- Clonality:Monoclonal
- Clone:1F3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 79
- Storage buffer:PBS, pH 7,2.
- Sequence:MVDHLLPVDENFSSPKCPVGYLGDRLVGRRAYHMLPSPVSEDDSDASSPCSCSSPDSQALCSCYGGGLGTESQDSILD*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: KLF15 (Kruppel-like factor 15)
Clonality: monoclonal
Clone: 1F3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human