Anti-KCNJ15 Mouse Monoclonal Antibody [clone: 1B2]
USBI128748
- Antibody type:Primary
- Antigen name:potassium inwardly-rectifying channel, subfamily J, member 15
- Antigen symbol:KCNJ15
- Clonality:Monoclonal
- Clone:1B2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:290 to 355
- Storage buffer:PBS, pH 7,2.
- Sequence:TSAVCQSRTSYIPEEIYWGFEFVPVVSLSKNGKYVADFSQFEQIRKSPDCTFYCADSEKQQLEEKY
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: KCNJ15 (potassium inwardly-rectifying channel, subfamily J, member 15)
Clonality: monoclonal
Clone: 1B2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human