Order Entry
Sweden
ContactUsLinkComponent
 
Anti-IFT81 Mouse Polyclonal Antibody
  USBI128326
Anti-IFT81 Mouse Polyclonal Antibody
  USBI128326

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    intraflagellar transport 81 homolog (Chlamydomonas)
  • Antigensymbol:
    IFT81
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Mouse
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Aminosyretal:
    1 to 676
  • Storage buffer:
    PBS, pH 7,2.
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Rening:
    purified by Protein A chromatography
  • Storlek:
    50 µg
  • Förp.:
    50 µG

 

 

Type: Primary
Antigen: IFT81 (intraflagellar transport 81 homolog (Chlamydomonas))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human