Anti-GTF2H4 Rabbit Polyclonal Antibody
Catalog # USBI127644
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:General transcription factor 2H, polypeptide 4, 52kDa
- Antigen symbol:GTF2H4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Format:Serum
- Form:liquid
- Amino acid number:1 to 462
- Sequence:MESTPSRGLNRVHLQCRNLQEFLGGLSPGVLDRLYGHPATCLAVFRELPSLAKNWVMRMLFLEQPLPQAAVALWVKKEFSKAQEESTGLLSGLRIWHTQLLPGGLQGLILNPIFRQNLRIALLGGGKAWSDDTSQLGPDKHARDVPSLDKYAEERWEVVLHFMVGSPSAAVSQDLAQLLSQAGLMKSTEPGEPPCITSAGFQFLLLDTPAQLWYFMLQYLQTAQSRGMDLVEILSFLFQLSFSTLGKDYSVEGMSDSLLNFLQHLREFGLVFQRKRKSRRYYPTRLAINLSSGVSGAGGTVHQPGFIVVETNYRLYAYTESELQIALIALFSEMLYRFPNMVVAQVTRESVQQAIASGITAQQIIHFLRTRAHPVMLKQTPVLPPTITDQIRLWELERDRLRFTEGVLYNQFLSQVDFELLLAHARELGVLVFENSAKRLMVVTPAGHSDVKRFWKRQKHSS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: GTF2H4 (general transcription factor IIH, polypeptide 4, 52kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human