Anti-NPSR1 Mouse Monoclonal Antibody [clone: 2F5]
Catalog # USBI127493
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Neuropeptide S Receptor 1
- Antigen symbol:NPSR1
- Clonality:Monoclonal
- Clone:2F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:2 to 54
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: NPSR1 (neuropeptide S receptor 1)
Clonality: monoclonal
Clone: 2F5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human