Anti-G Protein Coupled Receptor 48 Mouse Monoclonal Antibody [clone: 8F6]
USBI127056
- Antibody type:Primary
- Antigen name:G Protein Coupled Receptor 48
- Antigen symbol:GPR48
- Clonality:Monoclonal
- Clone:8F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:302 to 360
- Storage buffer:PBS, pH 7,2.
- Sequence:QGNLTVCDCCESFLLTKPVSCKHLIKSHSCPALAVASCQRPEGYWSDCGTQSAHSDYADEEDSFVSDSSDQVQACGRACFYQSRGFPLVRYAYNLPRVK
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: G Protein Coupled Receptor 48
Clonality: monoclonal
Clone: 8F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human