Anti-FZD7 Mouse Monoclonal Antibody [clone: 4D9]
Catalog # USBI127047
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Frizzled 7
- Antigen symbol:FZD7
- Clonality:Monoclonal
- Clone:4D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:155 to 254
- Storage buffer:PBS, pH 7,2.
- Sequence:GAGEICVGQNTSDGSGGPGGGPTAYPTAPYLPDLPFTALPPGASDGRGRPAFPFSCPRQLKVPPYLGYRFLGERDCGAPCEPGRANGLMYFKEEERRFA*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: FZD7 (frizzled class receptor 7)
Clonality: monoclonal
Clone: 4D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human