Anti-COX17 Mouse Monoclonal Antibody [clone: 4G2]
USBI125246
- Antibody type:Primary
- Antigen name:COX17 cytochrome c oxidase copper chaperone
- Antigen symbol:COX17
- Clonality:Monoclonal
- Clone:4G2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 63
- Storage buffer:PBS, pH 7,2.
- Sequence:MPGLVDSNPAPPESQEKKPLKPCCACPETKKARDACIIEKGEEHCGHLIEAHKECMRALGFKI
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: COX17 (COX17 cytochrome c oxidase copper chaperone)
Clonality: monoclonal
Clone: 4G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human