Order Entry
Sweden
ContactUsLinkComponent
Anti-CALM2 Mouse Monoclonal Antibody [clone: 3F4-G5]
Anti-CALM2 Mouse Monoclonal Antibody [clone: 3F4-G5]
Catalog # USBI124257
Supplier:  US Biological
Anti-CALM2 Mouse Monoclonal Antibody [clone: 3F4-G5]
Catalog # USBI124257
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    calmodulin 2 (phosphorylase kinase, delta)
  • Antigen symbol:
    CALM2
  • Clonality:
    Monoclonal
  • Clone:
    3F4-G5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    MADQLTEEQIAEFKEAFSLFDKDGDGTITTKELGTVMRSLGQNPTEAELQDMINEVDADGNGTIDFPEFLTMMARKMKDTDSEEEIREAFRVFDKDGNGYISAAELRHVMTNLGEKLTDEEVDEMIREADIDGDGQVNYEEFVQMMTAK
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: CALM2 (calmodulin 2 (phosphorylase kinase, delta))
Clonality: monoclonal
Clone: 3F4-G5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human