Order Entry
Sweden
ContactUsLinkComponent
Anti-H2B (ubiquitinated) Mouse Monoclonal Antibody [clone: NRO3]
Anti-H2B (ubiquitinated) Mouse Monoclonal Antibody [clone: NRO3]
  MEDMMM-0029-P
 :  MédiMabs
Anti-H2B (ubiquitinated) Mouse Monoclonal Antibody [clone: NRO3]
  MEDMMM-0029-P
 :  MédiMabs

 

  • Antibody type:
    Primary
  • Antigen name:
    Histone H2B
  • Antigen symbol:
    H2B
  • Clonality:
    Monoclonal
  • Clone:
    NRO3
  • Conjugation:
    Unconjugated
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Modification:
    ubiquitinated
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: H2B (Histone H2B)
Clonality: Monoclonal
Clone: NRO3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: