Order Entry
Sweden
ContactUsLinkComponent
 
Anti-THRAP3 Rabbit Polyclonal Antibody (Biotin)
  USBI042816-BIOTIN
Anti-THRAP3 Rabbit Polyclonal Antibody (Biotin)
  USBI042816-BIOTIN

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Thyroid Hormone Receptor Associated Protein 3
  • Antigensymbol:
    THRAP3
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Biotin
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Form:
    liquid
  • Aminosyretal:
    926 to 955
  • Storage buffer:
    PBS, pH 7,2
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Rening:
    purified by Protein A chromatography
  • Storlek:
    200 µl
  • Förp.:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: THRAP3 (thyroid hormone receptor associated protein 3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human