Order Entry
Sweden
ContactUsLinkComponent
 
Anti-TACO1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
  USBI042567-APC
Anti-TACO1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
  USBI042567-APC

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    translational activator of mitochondrially encoded cytochrome c oxidase I
  • Antigensymbol:
    TACO1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    N-terminal
  • Form:
    liquid
  • Aminosyretal:
    49 to 77
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Rening:
    purified by Protein A chromatography
  • Storlek:
    200 µl
  • Förp.:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: TACO1 (translational activator of mitochondrially encoded cytochrome c oxidase I)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human