Order Entry
Sweden
ContactUsLinkComponent
Anti-PDE8A Rabbit Polyclonal Antibody
Anti-PDE8A Rabbit Polyclonal Antibody
  USBI039812
Anti-PDE8A Rabbit Polyclonal Antibody
  USBI039812

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    phosphodiesterase 8A
  • Antigensymbol:
    PDE8A
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Epitop:
    internal
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide.
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Rening:
    purified by Protein A chromatography
  • Storlek:
    200 µl
  • Förp.:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: PDE8A (phosphodiesterase 8A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human