Anti-MYH7 Rabbit Polyclonal Antibody
Catalog # USBI038729
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:myosin, heavy chain 7, cardiac muscle, beta
- Antigen symbol:MYH7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:184 to 211
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: MYH7 (myosin, heavy chain 7, cardiac muscle, beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse