Anti-MARCH1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBI038192-APC
Leverantör: US Biological
Specifikationer
- Antibody type:Primary
- Antigen name:Membrane associated ring-CH-type finger 2
- Antigen symbol:MARCH2
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Antigen synonyms:RNF172|MARCH-II|HSPC240
- Amino acid number:145 to 174
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifikationer
Om denna produkt
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MARCH1 (membrane-associated ring finger (C3HC4) 1, E3 ubiquitin protein ligase)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse