Anti-KCTD10 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # USBI037355-AP
Supplier: US Biological
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:potassium channel tetramerization domain containing 10
- Antigensymbol:KCTD10
- Klonalitet:Polyclonal
- Konjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flödescytometri:Yes
- Värd:Rabbit
- Isotyp:IgG
- Reaktivitet:Human
- Western blot:Yes
- Epitop:C-terminal
- Form:liquid
- Aminosyretal:272 to 301
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Rening:purified by Protein A chromatography
- Storlek:200 µl
- Förp.:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: KCTD10 (potassium channel tetramerization domain containing 10)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human