Order Entry
Sweden
ContactUsLinkComponent
Anti-GCNT7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-GCNT7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  USBI035973-FITC
undefined
Anti-GCNT7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  USBI035973-FITC

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Glucosaminyl (N-acetyl) transferase 7
  • Antigensymbol:
    GCNT7
  • Klonalitet:
    Polyclonal
  • Konjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Form:
    liquid
  • Aminosyretal:
    323 to 351
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Rening:
    purified by Protein A chromatography
  • Storlek:
    200 µl
  • Förp.:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: GCNT7 (glucosaminyl (N-acetyl) transferase family member 7)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human