Anti-CPSF3L Rabbit Polyclonal Antibody (PE (Phycoerythrin))
USBI034179-PE
- Antibody type:Primary
- Antigen name:cleavage and polyadenylation specific factor 3-like
- Antigen symbol:CPSF3L
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:98 to 126
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50
Do not freeze!
Type: Primary
Antigen: CPSF3L (cleavage and polyadenylation specific factor 3-like)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human