Anti-COL17A1 Rabbit Polyclonal Antibody
USBI034083
- Antibody type:Primary
- Antigen name:collagen, type XVII, alpha 1
- Antigen symbol:COL17A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Amino acid number:475 to 504
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: COL17A1 (collagen, type XVII, alpha 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human