Order Entry
Sweden
ContactUsLinkComponent
Anti-CHST9 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anti-CHST9 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # USBI033889-PE
Supplier:  US Biological
undefined
Anti-CHST9 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # USBI033889-PE
Supplier:  US Biological

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9
  • Antigensymbol:
    CHST9
  • Klonalitet:
    Polyclonal
  • Konjugation:
    PE (Phycoerythrin)
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    N-terminal
  • Form:
    liquid
  • Aminosyretal:
    20 to 50
  • Storage buffer:
    PBS, pH 7,2
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Rening:
    purified by Protein A chromatography
  • Storlek:
    200 µl
  • Förp.:
    200 µl

Specifications

About this item

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: CHST9 (carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human